Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) isopropoxide, 98+%

Catalog No. AC194702500
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC194702500 250 mL Glass bottle
AC194700050 5 mL Glass bottle
AC194700010 1 L Glass bottle
AC194700025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC194702500 Supplier Thermo Scientific Chemicals Supplier No. 194702500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 546-68-9 | C12H28O4Ti | 284.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-68-9
Molecular Formula C12H28O4Ti
Molecular Weight (g/mol) 284.26
InChI Key VXUYXOFXAQZZMF-UHFFFAOYSA-N
Synonym titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
PubChem CID 11026
IUPAC Name propan-2-olate;titanium(4+)
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Specifications

Melting Point >15.0°C
Density 0.9500g/mL
Boiling Point 240.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 16.6 to 17.3% (Ti)
Packaging Glass bottle
Refractive Index 1.4654 to 1.4684
Linear Formula Ti[OCH(CH3)2]4
Quantity 250 mL
Beilstein 01,II,382
Fieser 11,92; 12,90; 13,13; 14,61; 15,308; 16,54; 17,347
Merck Index 15,9636
Specific Gravity 0.95
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
Viscosity 4.3 mPa.s (25°C)
Formula Weight 284.26
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Titanium(IV) isopropoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remov
EINECSNumber 208-909-6
Recommended Storage Product may darken during storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.