Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Chromate 99%, Thermo Scientific™

Catalog No. AC211250500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC211250500 50 g
AC211250100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC211250500 Supplier Thermo Scientific Chemicals Supplier No. 211250500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-01-2
Molecular Formula Ag2CrO4
Molecular Weight (g/mol) 331.73
InChI Key OJKANDGLELGDHV-UHFFFAOYSA-N

Specifications

Color Black to Gray
Physical Form Powder
Quantity 50 g
Linear Formula Ag2CrO4
Fieser 01,1005
Merck Index 15, 8649
SMILES [Ag+].[Ag+].[O-][Cr]([O-])(=O)=O
IUPAC Name disilver(1+) dioxochromiumbis(olate)
Formula Weight 331.72
Percent Purity ≥98.5% (AG2CRO4)
Density 5.62
Chemical Name or Material Silver chromate, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement LIGHT SENSITIVE
EINECSNumber 232-043-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.