Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylselenyl bromide, 98%

Catalog No. AC223060250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC223060250 25 g Glass bottle
AC223060050 5 g Glass bottle
2 options

Catalog No. AC223060250

Supplier: Thermo Scientific Chemicals 223060250

Only null left
Add to Cart

CAS: 34837-55-3 | C6H5BrSe | 235.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34837-55-3
Molecular Formula C6H5BrSe
Molecular Weight (g/mol) 235.97
MDL Number MFCD00000047
InChI Key LCEFEIBEOBPPSJ-UHFFFAOYSA-N
Synonym phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
PubChem CID 123446
IUPAC Name phenyl selenohypobromite
SMILES C1=CC=C(C=C1)[Se]Br

Specifications

Melting Point 60°C to 62°C
Color Black to Brown
Boiling Point 107°C to 108°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5SeBr
Quantity 25 g
Beilstein 06, II, 319
Fieser 06,459; 07,286; 11,33; 13,26; 01,1047; 11,487; 16,272
Solubility Information Solubility in water: insoluble
Formula Weight 235.97
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Phenylselenyl bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effec
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air
EINECSNumber 252-238-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.