Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone oxime, 98%

Catalog No. AC342330250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC342330250 25 g Glass bottle
AC342330050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC342330250 Supplier Thermo Scientific Chemicals Supplier No. 342330250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 574-66-3 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 574-66-3
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00051461
InChI Key DNYZBFWKVMKMRM-UHFFFAOYSA-N
Synonym benzophenone oxime, diphenylmethanone oxime, benzophenoxime, methanone, diphenyl-, oxime, diphenyl ketoxime, benzophenoneoxime, diphenylmethylene hydroxylamine, benzophenone, oxime, unii-2dlj8c37dq, diphenylketone oxime
PubChem CID 11324
IUPAC Name N-benzhydrylidenehydroxylamine
SMILES ON=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 140°C to 144°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CNOH(C6H5)
Quantity 25 g
Merck Index 15, 1100
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in hexane,soluble in benzene(warm),chloroform,,dichloromethane,ethyl acetate,methanol
Formula Weight 197.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzophenone oxime, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 209-373-6
RTECSNumber DJ1810000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.