Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orange G (Certified Biological Stain), Fisher Chemical

Catalog No. O26725 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O267-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. O267-25

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in histology

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00012457
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym Acid Orange G, 1-Phenylazo-2-naphthol-6, 8-disulfonic Acid Disodium Salt, 7-Hydroxy-8-(phenylazo)-1, 3-naphthalenedisulfonic Acid Sodium Salt
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 141°C
Bio. Stain Comm. Certification Pass Test
Dye Content Report %
Formula Weight 452.38
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 16230
Color Orange/Yellow
Quantity 25 g
Physical Form Solid
Chemical Name or Material Orange G
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.