Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aβ1-14-εK-HBsAg3 Th | 98.8% | 3893.45 | 5 MG
SDP

Catalog No. 5000399625
Encompass_Preferred

Aβ1-14-εK-HBsAg3 Th is a biologically active peptide, identified as one of UB-311's Aβ1-14-targeting peptides (B-cell epitope). It is for research use only. This peptide has a purity of 98.76% and a molecular weight of 3893.45.

  • Purity of 98.76%
  • Molecular weight of 3893.45
  • Chemical formula: C175H287N49O51
  • Appears as a solid
  • Color is white to off-white
  • Full sequence: Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-{εLys}-Lys-Lys-Lys-Ile-Ile-Thr-Ile-Thr-Arg-Ile-Ile-Thr-Ile-Ile-Thr-Thr-Ile-Asp
  • Shortened sequence: DAEFRHDSGYEVHH-{εLys}-KKK-IITITRIITIITTID

Catalog No. 50-003-99625 Supplier Medchemexpress LLC Supplier No. HYP53325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aβ1-14-εK-HBsAg3 Th is a biologically active peptide, identified as one of UB-311's Aβ1-14-targeting peptides (B-cell epitope). It is for research use only. This peptide has a purity of 98.76% and a molecular weight of 3893.45.

  • Purity of 98.76%
  • Molecular weight of 3893.45
  • Chemical formula: C175H287N49O51
  • Appears as a solid
  • Color is white to off-white
  • Full sequence: Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-{εLys}-Lys-Lys-Lys-Ile-Ile-Thr-Ile-Thr-Arg-Ile-Ile-Thr-Ile-Ile-Thr-Thr-Ile-Asp
  • Shortened sequence: DAEFRHDSGYEVHH-{εLys}-KKK-IITITRIITIITTID

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.