Learn More
Medchemexpress LLC Aβ1-42 aggregation inhibitor 1 | 2439107-75-0 | 468.66 | 1 ML
SDP
Catalog No. 5000360481
Encompass_Preferred
Aβ1-42 aggregation inhibitor 1 inhibits AChE (acetylcholinesterase) and BuChE (butyrylcholinesterase) with IC50 values of 2.64 μM and 1.29 μM, respectively. It also inhibits self-mediated Aβ1-42 aggregation by 51.29% at a concentration of 25 μM. This compound has potential for research into anti-Alzheimer's disease.
  • Inhibits AChE (acetylcholinesterase) with an IC50 of 2.64 μM.
  • Inhibits BuChE (butyrylcholinesterase) with an IC50 of 1.29 μM.
  • Inhibits self-mediated Aβ1-42 aggregation by 51.29% at 25 μM.
  • Has potential for anti-Alzheimer's disease research.
Catalog No. 50-003-60481 Supplier Medchemexpress LLC Supplier No. HY15000310MM/1ML
Aβ1-42 aggregation inhibitor 1 inhibits AChE (acetylcholinesterase) and BuChE (butyrylcholinesterase) with IC50 values of 2.64 μM and 1.29 μM, respectively. It also inhibits self-mediated Aβ1-42 aggregation by 51.29% at a concentration of 25 μM. This compound has potential for research into anti-Alzheimer's disease.
  • Inhibits AChE (acetylcholinesterase) with an IC50 of 2.64 μM.
  • Inhibits BuChE (butyrylcholinesterase) with an IC50 of 1.29 μM.
  • Inhibits self-mediated Aβ1-42 aggregation by 51.29% at 25 μM.
  • Has potential for anti-Alzheimer's disease research.