Learn More
Ace Glass, Inc. PTFE stir blade, 19x60mm, 10mm shafts
SDP
Catalog No. 509558246
Encompass_Preferred
19x60mm PTFE stir blade for 10mm O.D. button type stir shafts.
Catalog No. 50-955-8246 Supplier Ace Glass, Inc. Supplier No. 808507
19x60mm PTFE stir blade for 10mm O.D. button type stir shafts.