Learn More
TOKYO RIKAKIKAI CO LTD A-1000S ASPIRATOR VACUUM PUMP
Catalog No. 50855970
Encompass_Preferred
Pump, Aspirator, Eyela; A-1000S; Maximum Vacuum, 8.7mbar at 5 deg. C, 12.3mbar at 10 deg. C, 23.3mbar at 20 deg. C; PP tank is 10-1/8W x 14-1/4D x 8-1/2 in.H; Capacity, 10L
Catalog No. 50-855-970 Supplier TOKYO RIKAKIKAI CO LTD Supplier No. 210539
Pump, Aspirator, Eyela; A-1000S; Maximum Vacuum, 8.7mbar at 5 deg. C, 12.3mbar at 10 deg. C, 23.3mbar at 20 deg. C; PP tank is 10-1/8W x 14-1/4D x 8-1/2 in.H; Capacity, 10L