Learn More
Miele, Inc. A 102 Upper basket with spray
Catalog No. 502870800
Encompass_Preferred
A 102 Upper basket, with spray arm, height adjustable for inserts. Vertical clearance 205+/-30mm
Catalog No. 50-287-0800 Supplier Miele, Inc. Supplier No. 12645600
A 102 Upper basket, with spray arm, height adjustable for inserts. Vertical clearance 205+/-30mm