Learn More
TARGETMOL CHEMICALS INC A 1120 10MG
SDP
Catalog No. 502880506
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. A 1120 is a nonretinoid retinol-binding protein 4 (RBP4) antagonist (Ki 8.3 nM), which disrupts the interaction between RBP4 and its binding partner transthyretin. Purity 99.11%
Catalog No. 50-288-0506 Supplier TARGETMOL CHEMICALS INC Supplier No. T1020310MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. A 1120 is a nonretinoid retinol-binding protein 4 (RBP4) antagonist (Ki 8.3 nM), which disrupts the interaction between RBP4 and its binding partner transthyretin. Purity 99.11%