Learn More
Medchemexpress LLC A 1120 10mM 1mL | 1152782-19-8 | 10MM 1ML
SDP
Catalog No. 5000240100
Encompass_Preferred
A 1120 is a high-affinity nonretinoid retinol-binding protein 4 (RBP4) antagonist with a Ki value of 8 3 nM A 1120 disrupts the interaction between RBP4 and its binding partner transthyretin[1 [2
Catalog No. 50-002-40100 Supplier Medchemexpress LLC Supplier No. HY10763310MM 1ML
A 1120 is a high-affinity nonretinoid retinol-binding protein 4 (RBP4) antagonist with a Ki value of 8 3 nM A 1120 disrupts the interaction between RBP4 and its binding partner transthyretin[1 [2