Learn More
Medchemexpress LLC HY-107633 5mg Medchemexpress, A 1120 CAS:1152782-19-8 Purity:>98%
SDP
Catalog No. 501932291
Encompass_Preferred
Medchemexpress, HY-107633 5mg A 1120 CAS:1152782-19-8 A 1120, a nonretinoid retinol-binding protein 4 (RBP4) antagonist, has a high affinity with the K i value of 8.3 nM, which disrupts the interaction between RBP4 and its binding partner transthyretin [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2291 Supplier Medchemexpress LLC Supplier No. HY1076335MG
Medchemexpress, HY-107633 5mg A 1120 CAS:1152782-19-8 A 1120, a nonretinoid retinol-binding protein 4 (RBP4) antagonist, has a high affinity with the K i value of 8.3 nM, which disrupts the interaction between RBP4 and its binding partner transthyretin [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.