Learn More
Selleck Chemical LLC A-1155463 E2926-25MG
SDP
Catalog No. 5000040304
Encompass_Preferred
A-1155463 is a highly potent and selective BCL-XL inhibitor with EC50 of 65 nM in H146 cells
Catalog No. 50-000-40304 Supplier Selleck Chemical LLC Supplier No. E292625MG
A-1155463 is a highly potent and selective BCL-XL inhibitor with EC50 of 65 nM in H146 cells