Learn More
Selleck Chemical LLC A-1155463 5mg 1235034-55-5
SDP
Catalog No. NC1298858
Encompass_Preferred
A-1155463, a highly potent and selective BCL-XL inhibitor, shows picomolar binding affinity to BCL-XL, and >1000-fold weaker binding to BCL-2 and related proteins BCL-W(Ki=19 nM) and MCL-1(Ki>440 nM). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1298858 Supplier Selleck Chemical LLC Supplier No. S78005MG
A-1155463, a highly potent and selective BCL-XL inhibitor, shows picomolar binding affinity to BCL-XL, and >1000-fold weaker binding to BCL-2 and related proteins BCL-W(Ki=19 nM) and MCL-1(Ki>440 nM). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.