Learn More
Selleck Chemical LLC A-1155463 Dihydrochloride
SDP
Catalog No. 5000002878
Encompass_Preferred
A-1155463 Dihydrochloride a highly potent and selective BCL-XL inhibitor shows picomolar binding affinity to BCL-XL and 1000-fold weaker binding to BCL-2 and related proteins BCL-W(Ki 19 nM) and MCL-1(Ki 440 nM)
Catalog No. 50-000-02878 Supplier Selleck Chemical LLC Supplier No. S780025MG
A-1155463 Dihydrochloride a highly potent and selective BCL-XL inhibitor shows picomolar binding affinity to BCL-XL and 1000-fold weaker binding to BCL-2 and related proteins BCL-W(Ki 19 nM) and MCL-1(Ki 440 nM)