Learn More
Selleck Chemical LLC A-1210477
SDP
Supplier:  Selleck Chemical LLC S779025MG
Encompass_Preferred
A-1210477 is a potent and selective MCL-1 inhibitor with Ki and IC50 of 0 454 nM and 26 2 nM respectively 100-fold selectivity over other Bcl-2 family members
Catalog No. 50-000-00756