Learn More
Medchemexpress LLC A-1210477 | 1668553-26-1 | MFCD28399118 | 99.9% | 850.04 g/mol | C46H55N7O7S | 25MG
SDP
Catalog No. 5000236308
Encompass_Preferred
A-1210477 is a potent, selective small-molecule inhibitor of the anti-apoptotic protein MCL-1 used as a research tool to study apoptosis and MCL-1-dependent cancer biology. It binds MCL-1 with sub-nanomolar affinity and disrupts MCL-1 protein-protein interactions to promote apoptosis in sensitive cell lines.
  • Potent, selective MCL-1 inhibition with Ki ≈ 0.45 nM.
  • Disrupts MCL-1 interactions to induce programmed cell death.
  • Suitable for cell-based assays and mechanistic studies of apoptosis.
  • High purity for reproducible experimental results.
  • Solid powder formulation with recommended cold storage for stability.
Catalog No. 50-002-36308 Supplier Medchemexpress LLC Supplier No. HY1246825MG
A-1210477 is a potent, selective small-molecule inhibitor of the anti-apoptotic protein MCL-1 used as a research tool to study apoptosis and MCL-1-dependent cancer biology. It binds MCL-1 with sub-nanomolar affinity and disrupts MCL-1 protein-protein interactions to promote apoptosis in sensitive cell lines.
  • Potent, selective MCL-1 inhibition with Ki ≈ 0.45 nM.
  • Disrupts MCL-1 interactions to induce programmed cell death.
  • Suitable for cell-based assays and mechanistic studies of apoptosis.
  • High purity for reproducible experimental results.
  • Solid powder formulation with recommended cold storage for stability.