Learn More
TARGETMOL CHEMICALS INC A-1210477 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T263225MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. A-1210477 is an effective and specific MCL-1 and Bcl-2 inhibitor (Ki/IC50 0.454/26.2 nM). purity: 99%
Catalog No. 50-258-0708