Learn More
Medchemexpress LLC HY-19741 10mg Medchemexpress, A-1331852 CAS:1430844-80-6 Purity:>98%
SDP
Catalog No. 502029291
Encompass_Preferred
Medchemexpress, HY-19741 10mg A-1331852 CAS:1430844-80-6 A-1331852 is an orally available BCL-XL selective inhibitor with a Ki of less than 10 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9291 Supplier Medchemexpress LLC Supplier No. HY1974110MG
Medchemexpress, HY-19741 10mg A-1331852 CAS:1430844-80-6 A-1331852 is an orally available BCL-XL selective inhibitor with a Ki of less than 10 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.