Learn More
Miele, Inc. A 150 lower basket for modules
Catalog No. NC1126954
Encompass
A 150 Lower basket
For the optimum loading of injector modules or inserts.
Lower basket with two module connections
For holding up to 2 injector modules or inserts
Automatic closing of the connections when not in use
Clearances: width 19 4/9" (494 mm), length 17 5/16" (440 mm )
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC1126954 Supplier Miele, Inc. Supplier No. 09862400
A 150 Lower basket
For the optimum loading of injector modules or inserts.
Lower basket with two module connections
For holding up to 2 injector modules or inserts
Automatic closing of the connections when not in use
Clearances: width 19 4/9" (494 mm), length 17 5/16" (440 mm )
Visit www.mieleusa.com/professional for technical information.