Learn More
CPC Scientific H-Ala-Gly-Glu-Gly-Leu-Ser-Ser-Pro-Phe-Trp-Ser-Leu-Ala-Ala-Pro-Gln-Arg-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MMNP001B
Encompass
SEQUENCE: H-Ala-Gly-Glu-Gly-Leu-Ser-Ser-Pro-Phe-Trp-Ser-Leu-Ala-Ala-Pro-Gln-Arg-Phe-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: AGEGLSSPFWSLAAPQRF-NH2MOLECULAR FORMULA: C89H130N24O24MOLECULAR WEIGHT: 1920.16STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [99588-52-0]SYNONYMS: RESEARCH AREA: PainREFERENCES:N.A.Elshourbagy et al., J. Biol. Chem., 275, 25965 (2000)SKU(s): MMNP-001A, MMNP-001B
Catalog No. 50-210-9742