Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC A-192621 | 195529-54-5 | 99.2% | 1 ML
SDP

Catalog No. 5000234155
Encompass_Preferred

A-192621 is a potent, selective, nonpeptide endothelin B (ETB) receptor antagonist supplied for research use and commonly provided as a 10 mM solution in DMSO.

  • Potent, selective endothelin B receptor antagonist (IC50 = 4.5 nM; Ki = 8.8 nM).
  • Supplied as a ready-to-use 10 mM solution in DMSO for in vitro studies.
  • High purity (≈99.2%), suitable for biochemical and pharmacological assays.
  • Molecular weight 558.66 g/mol; formula C33H38N2O6.
  • Recommended formulations available for in vivo dosing (e.g., 10% DMSO/40% PEG300/5% Tween-80/45% saline).

Catalog No. 50-002-34155 Supplier Medchemexpress LLC Supplier No. HY12029510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A-192621 is a potent, selective, nonpeptide endothelin B (ETB) receptor antagonist supplied for research use and commonly provided as a 10 mM solution in DMSO.

  • Potent, selective endothelin B receptor antagonist (IC50 = 4.5 nM; Ki = 8.8 nM).
  • Supplied as a ready-to-use 10 mM solution in DMSO for in vitro studies.
  • High purity (≈99.2%), suitable for biochemical and pharmacological assays.
  • Molecular weight 558.66 g/mol; formula C33H38N2O6.
  • Recommended formulations available for in vivo dosing (e.g., 10% DMSO/40% PEG300/5% Tween-80/45% saline).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.