Learn More
Selleck Chemical LLC A-196 2mg 1982372-88-2
SDP
Supplier:  Selleck Chemical LLC S79832MG
Encompass_Preferred
A-196 is a potent and selective inhibitor of SUV420H1 and SUV420H2 with IC50 values of 0.025 and 0.144 ?M, respectively; more than 100-fold selective over other histone methyltransferases and non-epigenetic targets. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5509