Learn More
Vici Precision Sampling, Inc. A-2 GAS SYRINGE .25ML
Catalog No. NC9894931
Encompass_Preferred
Gas Syringe Series A-2,250 microliter(0.25 ml),Push-button valve allows instantaneous injection,Smaller volumes great for small volatile samples,allows sample storage to 250 psi, PTFE plunger tip, PTFE-sealed needle, and ultrasmooth bore
Catalog No. NC9894931 Supplier Vici Precision Sampling, Inc. Supplier No. 050031
Gas Syringe Series A-2,250 microliter(0.25 ml),Push-button valve allows instantaneous injection,Smaller volumes great for small volatile samples,allows sample storage to 250 psi, PTFE plunger tip, PTFE-sealed needle, and ultrasmooth bore