Learn More
Miele, Inc. A 200 Mobile unit for up to 38
Catalog No. 502870756
Encompass_Preferred
A 200 Mobile unit for up to 38 pipettes in 3 rows (excludes PG 8504)
Catalog No. 50-287-0756 Supplier Miele, Inc. Supplier No. 12667060
A 200 Mobile unit for up to 38 pipettes in 3 rows (excludes PG 8504)