Learn More
Miele, Inc. A 200 mobile units for pipettes
Catalog No. 501713001
Encompass_Preferred
A 200 Mobile unit
For the optimum loading of 38 pipettes.
For 10 100 ml pipettes in row 1, distance 20 mm
For 14 25 ml pipettes in a 2nd row, distance 26 mm
For 14 10 ml pipettes in a 3rd row, distance 26 mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3001 Supplier Miele, Inc. Supplier No. 09862420
A 200 Mobile unit
For the optimum loading of 38 pipettes.
For 10 100 ml pipettes in row 1, distance 20 mm
For 14 25 ml pipettes in a 2nd row, distance 26 mm
For 14 10 ml pipettes in a 3rd row, distance 26 mm
Visit www.mieleusa.com/professional for technical information.