Learn More
Selleck Chemical LLC A-205804-10mg
SDP
Catalog No. 502389625
Encompass_Preferred
A-205804 is a potent and selective inhibitor of E-selectin and ICAM-1 expression with IC50 of 20 nM and 25 nM respectively.
Catalog No. 50-238-9625 Supplier Selleck Chemical LLC Supplier No. S288510MG
A-205804 is a potent and selective inhibitor of E-selectin and ICAM-1 expression with IC50 of 20 nM and 25 nM respectively.