Learn More
TARGETMOL CHEMICALS INC A-205804 25MG
SDP
Catalog No. 502580946
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A-205804 is a specific and effective inhibitor of E-selectin( IC50=20 nM ) and ICAM-1(IC50=25 nM) expression. purity: 98%
Catalog No. 50-258-0946 Supplier TARGETMOL CHEMICALS INC Supplier No. T225425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A-205804 is a specific and effective inhibitor of E-selectin( IC50=20 nM ) and ICAM-1(IC50=25 nM) expression. purity: 98%