Learn More
TARGETMOL CHEMICALS INC A 21960 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T264085MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. A 21960 (Ethyl 2-chloroacetoacetate) inhibits synapse formation between resolvase and two directly repeated res sites blocks site-specific recombination reactions and can be used to study bacterial infections. purity: 99%
Catalog No. 50-258-0698