Learn More
Enzo Life Sciences Antibiotic A-23187 (Calcimycin) (10mg). CAS: 52665-69-7
SDP
Supplier:  Enzo Life Sciences BMLCA1000010
Encompass_Preferred
Antibiotic possessing weak in vitro antimicrobial activity against gram positive bacteria and fungi. A highly selective divalent cation ionophore, commonly used to increase intracellular Ca2+ levels in intact cells. Purity: ≥99% (TLC). Solubility: Soluble in DMSO an, 100% ethanol or methylene chloride (10mg/ml); slightly soluble in water. Solubilize in DMSO or ethanol prior to preparing aqueous solutions of very low concentrations. Long Term Storage: -20°C.
Catalog No. 50-201-0349