Learn More
Medchemexpress LLC A-286982 1mg | 280749-17-9 | 1MG
SDP
Catalog No. 5000240108
Encompass_Preferred
A-286982 is a potent and allosteric LFA-1/ICAM-1 interaction inhibitor with IC50s of 44 nM and 35 nM in an LFA-1/ICAM-1 binding and LFA-1-mediated cellular adhesion assay respectively[1 [2
Catalog No. 50-002-40108 Supplier Medchemexpress LLC Supplier No. HY1075871MG
A-286982 is a potent and allosteric LFA-1/ICAM-1 interaction inhibitor with IC50s of 44 nM and 35 nM in an LFA-1/ICAM-1 binding and LFA-1-mediated cellular adhesion assay respectively[1 [2