Learn More
Miele, Inc. A 300/2 module 2x4 (w/o nozzles)
Catalog No. 501713052
Encompass
A 300/2 Module
For optimum loading of laboratory glassware – can be equipped with 8 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Module w/o injector nozzles – equipped based on individual needs
For up to 8 injector nozzles
Part of the SimpleLoad system
Nozzle spacing 4 5/8" across the width, 4 5/8"across the depth
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3052 Supplier Miele, Inc. Supplier No. 11056770
A 300/2 Module
For optimum loading of laboratory glassware – can be equipped with 8 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Module w/o injector nozzles – equipped based on individual needs
For up to 8 injector nozzles
Part of the SimpleLoad system
Nozzle spacing 4 5/8" across the width, 4 5/8"across the depth
Visit www.mieleusa.com/professional for technical information.