Learn More
Miele, Inc. A 300/3 module 2x4 (with 4x A 840, 4x A 841)
Catalog No. 501713055
Encompass_Preferred
A 300/3 Module
For optimum loading of laboratory glassware – can be equipped with 8 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Equipped with 4 A 840 nozzles and 4 A 841 nozzles
SimpleLoad system
Nozzle spacing 4 5/8" across the width, 4 5/8"across the depth
Visit www.mieleusa.com/professional for technical information.
This item is not returnable. View return policy
Catalog No. 50-171-3055 Supplier Miele, Inc. Supplier No. 11116900
A 300/3 Module
For optimum loading of laboratory glassware – can be equipped with 8 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Equipped with 4 A 840 nozzles and 4 A 841 nozzles
SimpleLoad system
Nozzle spacing 4 5/8" across the width, 4 5/8"across the depth
Visit www.mieleusa.com/professional for technical information.