Learn More
Miele, Inc. A 301/4 module 3x6 (w/o nozzles)
Catalog No. 501713053
Encompass_Preferred
A 301/4 Module
For optimum loading of laboratory glassware – can be equipped with 18 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Module w/o injector nozzles – equipped based on individual needs
For up to 18 injector nozzles
Part of the SimpleLoad system
Nozzle spacing 6 3/16" across the width, 3 a"cross the depth
Visit www.mieleusa.com/professional for technical information.
This item is not returnable. View return policy
Catalog No. 50-171-3053 Supplier Miele, Inc. Supplier No. 11056780
A 301/4 Module
For optimum loading of laboratory glassware – can be equipped with 18 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Module w/o injector nozzles – equipped based on individual needs
For up to 18 injector nozzles
Part of the SimpleLoad system
Nozzle spacing 6 3/16" across the width, 3 a"cross the depth
Visit www.mieleusa.com/professional for technical information.