Learn More
Miele, Inc. A 301/5 module 3x6 (with 9x A 842, 9x A 843)
Catalog No. 501713056
Encompass_Preferred
A 301/5 Module
For optimum loading of laboratory glassware – can be equipped with 18 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Equipped with 9 A 842 nozzles and 9 A 843 nozzles
Part of the SimpleLoad system
Nozzle spacing 6 3/16" across the width, 3" across the depth
Visit www.mieleusa.com/professional for technical information.
This item is not returnable. View return policy
Catalog No. 50-171-3056 Supplier Miele, Inc. Supplier No. 11116950
A 301/5 Module
For optimum loading of laboratory glassware – can be equipped with 18 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Equipped with 9 A 842 nozzles and 9 A 843 nozzles
Part of the SimpleLoad system
Nozzle spacing 6 3/16" across the width, 3" across the depth
Visit www.mieleusa.com/professional for technical information.