Learn More
Miele, Inc. A 302/2 module 4x8 (w/o nozzles)
Catalog No. 501713054
Encompass_Preferred
A 302/2 Module
For optimum loading of laboratory glassware – can be equipped with 32 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Module w/o injector nozzles – equipped based on individual needs
For up to 32 injector nozzles
Part of the SimpleLoad system
Nozzle spacing 2 3/8" across the width, 2 1/4" across the depth
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3054 Supplier Miele, Inc. Supplier No. 11056790
A 302/2 Module
For optimum loading of laboratory glassware – can be equipped with 32 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Module w/o injector nozzles – equipped based on individual needs
For up to 32 injector nozzles
Part of the SimpleLoad system
Nozzle spacing 2 3/8" across the width, 2 1/4" across the depth
Visit www.mieleusa.com/professional for technical information.