Learn More
Miele, Inc. A 302/3 module 4x8 (with 16x A 844, 16x A 845)
Catalog No. 501713057
Encompass_Preferred
A 302/3 Module
For optimum loading of laboratory glassware – can be equipped with 32 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Equipped with 16 A 844 nozzles and 16 A 845 nozzles
Part of the SimpleLoad system
Nozzle spacing 2 3/8" across the width, 2 1/4" across the depth
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3057 Supplier Miele, Inc. Supplier No. 11116960
A 302/3 Module
For optimum loading of laboratory glassware – can be equipped with 32 nozzles.
Suitable for lab. glassware such as Erlenmeyer and round flasks.
Equipped with 16 A 844 nozzles and 16 A 845 nozzles
Part of the SimpleLoad system
Nozzle spacing 2 3/8" across the width, 2 1/4" across the depth
Visit www.mieleusa.com/professional for technical information.