Learn More
Miele, Inc. A 306 module for measuring cylinders
Catalog No. 501713004
Encompass_Preferred
A 306 Module
For the optimum loading of large volume laboratory glassware.
Suitable for 4 x 1 L graduated cylinder
Holds cylinders up to 19 ¾" long
Contact surfaces plastic coated
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3004 Supplier Miele, Inc. Supplier No. 11035210
A 306 Module
For the optimum loading of large volume laboratory glassware.
Suitable for 4 x 1 L graduated cylinder
Holds cylinders up to 19 ¾" long
Contact surfaces plastic coated
Visit www.mieleusa.com/professional for technical information.