Learn More
Miele, Inc. A 312 Module for up to 8 visco
Catalog No. 502870710
Encompass_Preferred
A 312 Module for up to 8 viscometers; includes 8 x A867 special nozzles
Catalog No. 50-287-0710 Supplier Miele, Inc. Supplier No. 11054470
A 312 Module for up to 8 viscometers; includes 8 x A867 special nozzles