Learn More
Apexbio Technology LLC A-317491(Synonyms: A317491, A-317491 sodium salt hydrate, P2X3 receptor antagonist A-317491, A 317491), 5mg, CAS: 475205-49-3.
SDP
Catalog No. 502240213
Encompass_Preferred
A-317491 (CAS 475205-49-3) is a selective high-affinity antagonist targeting the P2X3 and heteromeric P2X2/3 receptors ligand-gated ion channels responsive to ATP primarily located on sensory neurons involved in pain signaling It demonstrates potent inhibition with reported Ki values of 9 nM (P2X2/3) and 22 nM (P2X3) In rat dorsal root ganglion neurons A-317491 suppresses receptor-mediated currents (IC50 15 nM) in a dose-dependent manner In animal models intrathecal or peripheral administration attenuates hyperalgesia associated with inflammation and neuropathic pain underscoring its utility in pain research
This item is not returnable. View return policy
Catalog No. 50-224-0213 Supplier Apexbio Technology LLC Supplier No. A31345
A-317491 (CAS 475205-49-3) is a selective high-affinity antagonist targeting the P2X3 and heteromeric P2X2/3 receptors ligand-gated ion channels responsive to ATP primarily located on sensory neurons involved in pain signaling It demonstrates potent inhibition with reported Ki values of 9 nM (P2X2/3) and 22 nM (P2X3) In rat dorsal root ganglion neurons A-317491 suppresses receptor-mediated currents (IC50 15 nM) in a dose-dependent manner In animal models intrathecal or peripheral administration attenuates hyperalgesia associated with inflammation and neuropathic pain underscoring its utility in pain research