Learn More
Selleck Chemical LLC A-317491
SDP
Catalog No. 5000000761
Encompass_Preferred
A-317491 is a novel potent and selective non-nucleotide antagonist of P2X3 and P2X2/3 receptors with Ki values of 22 nM and 9 nM for human P2X3 and P2X2/3 receptors
Catalog No. 50-000-00761 Supplier Selleck Chemical LLC Supplier No. S851925MG
A-317491 is a novel potent and selective non-nucleotide antagonist of P2X3 and P2X2/3 receptors with Ki values of 22 nM and 9 nM for human P2X3 and P2X2/3 receptors