Learn More
Medchemexpress LLC A-350619 hydrochloride | 1217201-17-6 | ≥99.0% | 425.41 g/mol | C21H25ClN2OS·HCl | 10 MG
SDP
Catalog No. 5000240380
Encompass_Preferred
A-350619 hydrochloride is a small-molecule soluble guanylate cyclase (sGC) activator used in preclinical research. It enhances cGMP production by activating basal sGC and synergizing with nitric oxide, producing vasodilatory effects such as cavernosum smooth muscle relaxation and increased blood flow in animal models. The material is supplied as a hydrochloride salt for improved solubility.
  • High purity suitable for in vitro and in vivo studies.
  • Soluble hydrochloride salt with good DMSO and aqueous solubility.
  • Activates soluble guanylate cyclase and enhances NO-mediated signaling.
  • Demonstrated vasodilatory and penile erection effects in rodent models.
  • Store at 4°C to maintain stability.
Catalog No. 50-002-40380 Supplier Medchemexpress LLC Supplier No. HY10754810MG
A-350619 hydrochloride is a small-molecule soluble guanylate cyclase (sGC) activator used in preclinical research. It enhances cGMP production by activating basal sGC and synergizing with nitric oxide, producing vasodilatory effects such as cavernosum smooth muscle relaxation and increased blood flow in animal models. The material is supplied as a hydrochloride salt for improved solubility.
  • High purity suitable for in vitro and in vivo studies.
  • Soluble hydrochloride salt with good DMSO and aqueous solubility.
  • Activates soluble guanylate cyclase and enhances NO-mediated signaling.
  • Demonstrated vasodilatory and penile erection effects in rodent models.
  • Store at 4°C to maintain stability.