Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC A-381393 10mM 1mL | 726174-00-1 | 320.43 g·mol-1 | C20H24N4 | 1 ML
SDP

Catalog No. 5000307737
Encompass_Preferred

A-381393 is a potent, selective, brain-penetrant dopamine D4 receptor antagonist supplied as a 10 mM solution in DMSO for laboratory research applications (research use only). It exhibits low-nanomolar affinity for D4 subtypes and high selectivity over other dopamine receptors.

  • Supplied as a 10 mM solution in DMSO.
  • Packaged in a 1 mL vial for convenient dosing.
  • Molecular weight 320.43 g·mol-1; formula C20H24N4.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 2 years) or -20°C (up to 1 year).
  • Intended for in vitro pharmacology and neuronal signaling research; not for human use.

Catalog No. 50-003-07737 Supplier Medchemexpress LLC Supplier No. HY11694110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A-381393 is a potent, selective, brain-penetrant dopamine D4 receptor antagonist supplied as a 10 mM solution in DMSO for laboratory research applications (research use only). It exhibits low-nanomolar affinity for D4 subtypes and high selectivity over other dopamine receptors.

  • Supplied as a 10 mM solution in DMSO.
  • Packaged in a 1 mL vial for convenient dosing.
  • Molecular weight 320.43 g·mol-1; formula C20H24N4.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 2 years) or -20°C (up to 1 year).
  • Intended for in vitro pharmacology and neuronal signaling research; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.