Learn More
Medchemexpress LLC HY-101512 10mg Medchemexpress, A-395 CAS:2089148-72-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10151210MG
Encompass_Preferred
Medchemexpress, HY-101512 10mg A-395 CAS:2089148-72-9 A-395 is an antagonist of polycomb repressive complex 2 (PRC2) protein-protein interactions that potently inhibits the trimeric PRC2 complex (EZH2-EED-SUZ12) with an IC50 of 18 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7854