Learn More
Medchemexpress LLC A 410099.1, amine-Boc | 1613552-03-6 | 583.76 | 25 MG
SDP
Catalog No. 5000351158
Encompass_Preferred
A 410099.1, amine-Boc is a functionalized IAP ligand that can be utilized for synthesizing PROTACs, specifically PROTACs targeting BTK (PROTACs 4 and 5).
  • Functionalized IAP ligand.
  • Can be used for the synthesis of PROTACs.
  • Applicable for PROTACs targeting BTK.
  • For research use only.
Catalog No. 50-003-51158 Supplier Medchemexpress LLC Supplier No. 25MGHY136269
A 410099.1, amine-Boc is a functionalized IAP ligand that can be utilized for synthesizing PROTACs, specifically PROTACs targeting BTK (PROTACs 4 and 5).
  • Functionalized IAP ligand.
  • Can be used for the synthesis of PROTACs.
  • Applicable for PROTACs targeting BTK.
  • For research use only.