Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC A 410099.1, amine-Boc hydrochloride | 2374122-37-7 | 99.7% | C32H50ClN5O5 | 10MG
SDP

Catalog No. 5000259187
Encompass_Preferred

A 410099.1, amine-Boc hydrochloride is a Boc-protected amine hydrochloride derivative used as a functionalized IAP ligand and building block in PROTAC synthesis, including applications targeting Bruton's tyrosine kinase (BTK). Supplied for research use, it is characterized for solubility in DMSO and requires appropriate storage to maintain stability.

  • High purity: 99.7% (reported)
  • Boc-protected amine with an accessible amine functional handle for conjugation
  • White to off-white solid suitable for synthetic applications
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance may be required
  • Recommended storage: protect from light; use cold storage for solutions
  • Available in small mg quantities for research-scale synthesis

Catalog No. 50-002-59187 Supplier Medchemexpress LLC Supplier No. HY13626910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 410099.1, amine-Boc hydrochloride is a Boc-protected amine hydrochloride derivative used as a functionalized IAP ligand and building block in PROTAC synthesis, including applications targeting Bruton's tyrosine kinase (BTK). Supplied for research use, it is characterized for solubility in DMSO and requires appropriate storage to maintain stability.

  • High purity: 99.7% (reported)
  • Boc-protected amine with an accessible amine functional handle for conjugation
  • White to off-white solid suitable for synthetic applications
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance may be required
  • Recommended storage: protect from light; use cold storage for solutions
  • Available in small mg quantities for research-scale synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.