Learn More
Medchemexpress LLC Aβ42-in-2 | 1914989-80-2 | 98.0% | 430.50 g/mol | C24H26N6O2 | 5 MG
SDP
Catalog No. 5000223426
Encompass_Preferred
Aβ42-in-2 is a γ-secretase modulator reported to potently inhibit Aβ42 production (IC50 6.5 nM). Supplied as an off-white to light yellow solid for research use, the compound has a molecular weight of 430.50 g/mol and a reported purity of 98.0%.
  • Potent inhibitor of Aβ42 with an IC50 of 6.5 nM.
  • Reported purity of 98.0% for reliable experimental consistency.
  • Solid, off-white to light yellow appearance for straightforward handling.
  • Available in small quantities and as 10 mM DMSO solutions for assay readiness.
  • Storage conditions provided to maintain stability (4°C; in solvent -80°C/-20°C guidance).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-23426 Supplier Medchemexpress LLC Supplier No. HY1368665MG
Aβ42-in-2 is a γ-secretase modulator reported to potently inhibit Aβ42 production (IC50 6.5 nM). Supplied as an off-white to light yellow solid for research use, the compound has a molecular weight of 430.50 g/mol and a reported purity of 98.0%.
  • Potent inhibitor of Aβ42 with an IC50 of 6.5 nM.
  • Reported purity of 98.0% for reliable experimental consistency.
  • Solid, off-white to light yellow appearance for straightforward handling.
  • Available in small quantities and as 10 mM DMSO solutions for assay readiness.
  • Storage conditions provided to maintain stability (4°C; in solvent -80°C/-20°C guidance).
  • For research use only; not for human or clinical use.