Learn More
TARGETMOL CHEMICALS INC A-437203 5MG
SDP
Catalog No. 502581748
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A-437203 (Lu201640) is A kind of based on 1 h - pyrimidin - 2 - one sca ff old selective new powerful d3 receptor antagonist on D2 d3 and D4 receptor Ki value 71 respectively 1.6 and 6220 nM. purity: 98%
Catalog No. 50-258-1748 Supplier TARGETMOL CHEMICALS INC Supplier No. T102135MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A-437203 (Lu201640) is A kind of based on 1 h - pyrimidin - 2 - one sca ff old selective new powerful d3 receptor antagonist on D2 d3 and D4 receptor Ki value 71 respectively 1.6 and 6220 nM. purity: 98%