Learn More
Medchemexpress LLC A 438079 | 899507-36-9 | C13H9Cl2N5 | 100 MG
SDP
Catalog No. 5000376085
Encompass_Preferred
A 438079 is a potent and selective P2X7 receptor antagonist with a pIC50 of 6.9. It is identified by CAS number 899507-36-9 and chemical formula C13H9Cl2N5.
  • Potent and selective P2X7 receptor antagonist
  • Blocks BzATP-evoked changes in intracellular calcium concentrations
  • Reduces noxious and innocuous evoked activity of spinal neurons in neuropathic rats
  • Raises withdrawal thresholds in SNL and CCI models
  • Reduces seizure severity and neuronal death within the hippocampus
  • Partially prevents 6-OHDA-induced depletion of striatal DA stores
  • Reduces nociceptive behavior in the HC model
  • Provided as a white to off-white solid with 99.88% purity
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-003-76085 Supplier Medchemexpress LLC Supplier No. HY15488100MG
A 438079 is a potent and selective P2X7 receptor antagonist with a pIC50 of 6.9. It is identified by CAS number 899507-36-9 and chemical formula C13H9Cl2N5.
  • Potent and selective P2X7 receptor antagonist
  • Blocks BzATP-evoked changes in intracellular calcium concentrations
  • Reduces noxious and innocuous evoked activity of spinal neurons in neuropathic rats
  • Raises withdrawal thresholds in SNL and CCI models
  • Reduces seizure severity and neuronal death within the hippocampus
  • Partially prevents 6-OHDA-induced depletion of striatal DA stores
  • Reduces nociceptive behavior in the HC model
  • Provided as a white to off-white solid with 99.88% purity
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 2 years or -20°C for 1 year